3-(Methylsulfonyl)anilin

Lieferant: SIA ENAMINE

EN300-14687.5G
ENAMEN300-14687.5GEA 0 EUR
ENAMEN300-14687.5G
3-(Methylsulfonyl)anilin
3-(Methylsulfonyl)anilin
Formel: C₇H₉NO₂S
MDL: MFCD03764428
CAS-Nummer: 35216-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR