Methyl-4-(chlorsulfonyl)benzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 4-chlorosulphonylbenzoate

EN300-15237.1G
ENAMEN300-15237.1GEA 0 EUR
ENAMEN300-15237.1G
Methyl-4-(chlorsulfonyl)benzoat
Methyl-4-(chlorsulfonyl)benzoat
Formel: C₈H₇ClO₄S
Molecular Weight: 234.6568 g/mol
MDL: MFCD00627554
CAS-Nummer: 69812-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR