2-Methylimidazo[1,2-a]pyridin

Lieferant: SIA ENAMINE

EN300-18552
ENAMEN300-18552EA 0 EUR
ENAMEN300-18552
2-Methylimidazo[1,2-a]pyridin
2-Methylimidazo[1,2-a]pyridin
Formel: C₈H₈N₂
MDL: MFCD02251330
CAS-Nummer: 934-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR