2-Cyanphenylisothiocyanat

Lieferant: SIA ENAMINE

EN300-23803.1G
ENAMEN300-23803.1GEA 0 EUR
ENAMEN300-23803.1G
2-Cyanphenylisothiocyanat
2-Cyanphenylisothiocyanat
Formel: C₈H₄N₂S
MDL: MFCD00060361
CAS-Nummer: 81431-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR