3-(Brommethyl)-4-fluorbenzonitril

Lieferant: SIA ENAMINE

EN300-25341.5G
ENAMEN300-25341.5GEA 0 EUR
ENAMEN300-25341.5G
3-(Brommethyl)-4-fluorbenzonitril
3-(Brommethyl)-4-fluorbenzonitril
Formel: C₈H₅BrFN
CAS-Nummer: 856935-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR