(2-Methyl-1,3-thiazol-4-yl)methanol

Lieferant: SIA ENAMINE

Synonyme: (2-Methylthiazol-4-yl)methanol

EN300-27687.1G
ENAMEN300-27687.1GEA 0 EUR
ENAMEN300-27687.1G
(2-Methyl-1,3-thiazol-4-yl)methanol
(2-Methyl-1,3-thiazol-4-yl)methanol
Formel: C₅H₇NOS
Molecular Weight: 129.1802 g/mol
MDL: MFCD08703637
CAS-Nummer: 76632-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR