2-Amino-3-methylbenzamid

Lieferant: SIA ENAMINE

EN300-29544.1G
ENAMEN300-29544.1GEA 0 EUR
ENAMEN300-29544.1G
2-Amino-3-methylbenzamid
2-Amino-3-methylbenzamid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,1800 g/mol
MDL: MFCD04034517
CAS-Nummer: 1885-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR