Diethyl-aminomalonat Hydrochlorid

Lieferant: SIA ENAMINE

EN300-30329.5G
ENAMEN300-30329.5GEA 0 EUR
ENAMEN300-30329.5G
Diethyl-aminomalonat Hydrochlorid
Diethyl-aminomalonat Hydrochlorid
Formel: C₇H₁₄ClNO₄
MDL: MFCD00012510
CAS-Nummer: 13433-00-6
EINECS: 236-556-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR