2-Amino-6-methylbenzamid

Lieferant: SIA ENAMINE

EN300-30883.1G
ENAMEN300-30883.1GEA 0 EUR
ENAMEN300-30883.1G
2-Amino-6-methylbenzamid
2-Amino-6-methylbenzamid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
MDL: MFCD09729171
CAS-Nummer: 1885-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR