2-Chlor-6-nitrochinolin

Lieferant: SIA ENAMINE

EN300-39072.10
ENAMEN300-39072.10EA 0 EUR
ENAMEN300-39072.10
2-Chlor-6-nitrochinolin
2-Chloro-6-nitroquinoline
Formel: C₉H₅ClN₂O₂
Molecular Weight: 208,6000 g/mol
MDL: MFCD09040696
CAS-Nummer: 29969-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR