Monomethylterephthalat

Lieferant: SIA ENAMINE

Synonyme: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

EN300-41770
ENAMEN300-41770EA 0 EUR
ENAMEN300-41770
Monomethylterephthalat
Monomethylterephthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00002557
CAS-Nummer: 1679-64-7
EINECS: 216-849-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR