5-Nitro-2-picolin

Lieferant: SIA ENAMINE

Synonyme: 2-Methyl-5-nitropyridine

EN300-42381.5G
ENAMEN300-42381.5GEA 0 EUR
ENAMEN300-42381.5G
5-Nitro-2-picolin
5-Nitro-2-picolin
Formel: C₆H₆N₂O₂
Molecular Weight: 138.12 g/mol
MDL: MFCD04114179
CAS-Nummer: 21203-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR