Methyl-2-amino-5-methylbenzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 2-amino-5-methylbenzoate

EN300-42954.1G
ENAMEN300-42954.1GEA 0 EUR
ENAMEN300-42954.1G
Methyl-2-amino-5-methylbenzoat
Methyl-2-amino-5-methylbenzoat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,1900 g/mol
MDL: MFCD08234606
CAS-Nummer: 18595-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR