4-Mercaptobenzonitril

Lieferant: SIA ENAMINE

EN300-45156
ENAMEN300-45156EA 0 EUR
ENAMEN300-45156
4-Mercaptobenzonitril
4-Mercaptobenzonitril
Formel: C₇H₅NS
MDL: MFCD00272354
CAS-Nummer: 36801-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR