3-Amino-4-methylpyridin

Lieferant: SIA ENAMINE

EN300-51191.1G
ENAMEN300-51191.1GEA 0 EUR
ENAMEN300-51191.1G
3-Amino-4-methylpyridin
3-Amino-4-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108,14 g/mol
MDL: MFCD00128871
CAS-Nummer: 3430-27-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR