2-Hydroxy-5-(trifluormethyl)anilin

Lieferant: SIA ENAMINE

EN300-52162.1G
ENAMEN300-52162.1GEA 0 EUR
ENAMEN300-52162.1G
2-Hydroxy-5-(trifluormethyl)anilin
2-Hydroxy-5-(trifluormethyl)anilin
Formel: C₇H₆F₃NO
MDL: MFCD01731750
CAS-Nummer: 454-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR