2-Chlor-5-hydroxypyridin

Lieferant: SIA ENAMINE
Achtung

EN300-52976.5G
ENAMEN300-52976.5GEA 0 EUR
ENAMEN300-52976.5G
2-Chlor-5-hydroxypyridin
Formel: C₅H₄ClNO
MDL: MFCD00234050
CAS-Nummer: 41288-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR