4-Chlor-7-methylisatin

Lieferant: SIA ENAMINE

EN300-60309 EN300-60309.10
ENAMEN300-60309EA 0 EUR
ENAMEN300-60309 ENAMEN300-60309.10
4-Chlor-7-methylisatin
4-Chlor-7-methylisatin
Formel: C₉H₆ClNO₂
MDL: MFCD00047216
CAS-Nummer: 61258-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR