Methyl-3-methyl-5-nitrobenzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 3-methyl-5-nitrobenzoate

EN300-65558.1G
ENAMEN300-65558.1GEA 0 EUR
ENAMEN300-65558.1G
Methyl-3-methyl-5-nitrobenzoat
Methyl-3-methyl-5-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,1700 g/mol
MDL: MFCD12172988
CAS-Nummer: 482311-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR