Methyl-3-(hydroxymethyl)benzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 3-(hydroxymethyl)benzoate

EN300-65946.2
ENAMEN300-65946.2EA 0 EUR
ENAMEN300-65946.2
Methyl-3-(hydroxymethyl)benzoat
Methyl-3-(hydroxymethyl)benzoat
Formel: C₉H₁₀O₃
Molecular Weight: 166,1739 g/mol
MDL: MFCD06202752
CAS-Nummer: 67853-03-6
EINECS: 230-017-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR