Ethyl-2-amino-4-methyl-5-oxazolcarboxylat

Lieferant: SIA ENAMINE

Synonyme: Ethyl 2-amino-4-methyloxazole-5-carboxylate

EN300-75938.5G
ENAMEN300-75938.5GEA 0 EUR
ENAMEN300-75938.5G
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat
Ethyl-2-amino-4-methyl-5-oxazolcarboxylat
Formel: C₇H₁₀N₂O₃
Molecular Weight: 170,1659 g/mol
MDL: MFCD09028430
CAS-Nummer: 79221-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR