7-Brom-4-chlorchinolin

Lieferant: SIA ENAMINE

EN300-79433.1G
ENAMEN300-79433.1GEA 0 EUR
ENAMEN300-79433.1G
7-Brom-4-chlorchinolin
7-Bromo-4-chloroquinoline
Formel: C₉H₅BrClN
MDL: MFCD02684221
CAS-Nummer: 75090-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR