2-Brom-4-nitrobenzylalkohol

Lieferant: SIA ENAMINE

EN300-82746.1G
ENAMEN300-82746.1GEA 0 EUR
ENAMEN300-82746.1G
2-Brom-4-nitrobenzylalkohol
2-Brom-4-nitrobenzylalkohol
Formel: C₇H₆BrNO₃
MDL: MFCD12152256
CAS-Nummer: 183111-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR