2-Chlor-5-iodanilin

Lieferant: SIA ENAMINE

EN300-88616.5G
ENAMEN300-88616.5GEA 0 EUR
ENAMEN300-88616.5G
2-Chlor-5-iodanilin
2-Chlor-5-iodanilin
Formel: C₆H₅ClIN
MDL: MFCD11505641
CAS-Nummer: 16604-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR