1-Brom-2,3-difluor-5-nitrobenzol

Lieferant: SIA ENAMINE

Synonyme: 3-Brom-4,5-difluornitrobenzol

EN300-91854.10
ENAMEN300-91854.10EA 0 EUR
ENAMEN300-91854.10
1-Brom-2,3-difluor-5-nitrobenzol
1-Bromo-2,3-difluoro-5-nitrobenzene
Formel: C₆H₂BrF₂NO₂
MDL: MFCD03095339
CAS-Nummer: 374633-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR