3-(3-Methylphenoxy)propionsäure
Lieferant: SIA ENAMINE
EN30000043.1G
ENAMEN30000043.1GEA
0
EUR
ENAMEN30000043.1G
3-(3-Methylphenoxy)propionsäure
3-(3-Methylphenoxy)propionsäure
Formel:
C₁₀H₁₂O₃ |
MDL:
MFCD00642411 CAS-Nummer: 25173-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...