(3,5-Xylyloxy)essigsäure

Lieferant: SIA ENAMINE

Synonyme: (3,5-Dimethylphenoxy)essigsäure

EN30000070.1G
ENAMEN30000070.1GEA 0 EUR
ENAMEN30000070.1G
(3,5-Xylyloxy)essigsäure
(3,5-Xylyloxy)essigsäure
Formel: (C₆H₃(CH₃)₂)OCH₂COOH
Molecular Weight: 180,2 g/mol
MDL: MFCD00021755
CAS-Nummer: 5406-14-4
EINECS: 226-462-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR