3-(4-Methoxyphenoxy)propionsäure

Lieferant: SIA ENAMINE

Synonyme: 3-(4-Methoxyphenoxy)propanoic acid

EN30000079.1G
ENAMEN30000079.1GEA 0 EUR
ENAMEN30000079.1G
3-(4-Methoxyphenoxy)propionsäure
3-(4-Methoxyphenoxy)propionsäure
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00020515
CAS-Nummer: 20811-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR