3-(2-Methylphenoxy)propionsäure

Lieferant: SIA ENAMINE

EN30000080.1G
ENAMEN30000080.1GEA 0 EUR
ENAMEN30000080.1G
3-(2-Methylphenoxy)propionsäure
3-(2-Methylphenoxy)propionsäure
Formel: C₁₀H₁₂O₃
MDL: MFCD00642410
CAS-Nummer: 25173-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR