3-(2-Fluorphenoxy)propionsäure

Lieferant: SIA ENAMINE

EN30000103.1G
ENAMEN30000103.1GEA 0 EUR
ENAMEN30000103.1G
3-(2-Fluorphenoxy)propionsäure
3-(2-Fluorphenoxy)propionsäure
Formel: C₉H₉FO₃
MDL: MFCD01569309
CAS-Nummer: 2967-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR