3-(2-Methoxyphenoxy)propionsäure

Lieferant: SIA ENAMINE

EN30000121.1G
ENAMEN30000121.1GEA 0 EUR
ENAMEN30000121.1G
3-(2-Methoxyphenoxy)propionsäure
3-(2-Methoxyphenoxy)propionsäure
Formel: C₁₀H₁₂O₄
MDL: MFCD01931914
CAS-Nummer: 20370-80-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR