1-(Phenylsulfonyl)-4-piperidincarbonsäure

Lieferant: SIA ENAMINE

EN30000358.1G
ENAMEN30000358.1GEA 0 EUR
ENAMEN30000358.1G
1-(Phenylsulfonyl)-4-piperidincarbonsäure
1-(Phenylsulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₅NO₄S
MDL: MFCD00723116
CAS-Nummer: 122891-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR