1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: SIA ENAMINE

EN30000366.1G
ENAMEN30000366.1GEA 0 EUR
ENAMEN30000366.1G
1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Bromphenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄BrNO₄S
MDL: MFCD02331440
CAS-Nummer: 203519-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR