1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: SIA ENAMINE

EN30000368.1G
ENAMEN30000368.1GEA 0 EUR
ENAMEN30000368.1G
1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄N₂O₆S
MDL: MFCD01045658
CAS-Nummer: 331828-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR