(S)-1-(Phenylsulfonyl)-2-piperidincarbonsäure

Lieferant: SIA ENAMINE

Synonyme: (S)-1-(Phenylsulphonyl)pipecolinic acid

EN30000653.1G
ENAMEN30000653.1GEA 0 EUR
ENAMEN30000653.1G
(S)-1-(Phenylsulfonyl)-2-piperidincarbonsäure
(S)-1-(Phenylsulfonyl)-2-piperidincarbonsäure
Formel: C₁₂H₁₅NO₄S
Molecular Weight: 269,32 g/mol
MDL: MFCD02655853
CAS-Nummer: 223562-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR