5-Brom-2-thenoesäure

Lieferant: SIA ENAMINE

Synonyme: 5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure

EN30000712.1G
ENAMEN30000712.1GEA 0 EUR
ENAMEN30000712.1G
5-Brom-2-thenoesäure
5-Brom-2-thenoesäure
Formel: C₅H₃BrO₂S
Molecular Weight: 207,05 g/mol
MDL: MFCD00079725
CAS-Nummer: 7311-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR