Imidazo[1,2-a]pyridin-2-carbonsäure

Lieferant: SIA ENAMINE

EN30000834.1G
ENAMEN30000834.1GEA 0 EUR
ENAMEN30000834.1G
Imidazo[1,2-a]pyridin-2-carbonsäure
Imidazo[1,2-a]pyridin-2-carbonsäure
Formel: C₈H₆N₂O₂
MDL: MFCD03419462
CAS-Nummer: 64951-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR