4-Chlorpicolinsäure

Lieferant: SIA ENAMINE

Synonyme: 4-Chloropyridine-2-carboxylic acid

EN30000872.1G
ENAMEN30000872.1GEA 0 EUR
ENAMEN30000872.1G
4-Chlorpicolinsäure
4-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00191400
CAS-Nummer: 5470-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR