2-Methylimidazo[1,2-a]pyridin-3-carbaldehyd

Lieferant: SIA ENAMINE

EN30000962.1G
ENAMEN30000962.1GEA 0 EUR
ENAMEN30000962.1G
2-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
2-Methylimidazo[1,2-a]pyridin-3-carbaldehyd
Formel: C₉H₈N₂O
MDL: MFCD02251327
CAS-Nummer: 30384-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR