4'-(Difluormethoxy)acetophenon

Lieferant: SIA ENAMINE

EN30001080.1G
ENAMEN30001080.1GEA 0 EUR
ENAMEN30001080.1G
4'-(Difluormethoxy)acetophenon
4'-(Difluormethoxy)acetophenon
Formel: C₉H₈F₂O₂
MDL: MFCD00042250
CAS-Nummer: 83882-67-1
EINECS: 281-174-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR