5-Chlor-3-nitrosalicylaldehyd

Lieferant: SIA ENAMINE

EN30001100.1G
ENAMEN30001100.1GEA 0 EUR
ENAMEN30001100.1G
5-Chlor-3-nitrosalicylaldehyd
5-Chlor-3-nitrosalicylaldehyd
Formel: C₇H₄ClNO₄
MDL: MFCD00462959
CAS-Nummer: 16634-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR