7-Methylisatin

Lieferant: SIA ENAMINE

Synonyme: 7-Methylindoline-2,3-dione

EN30001162.1G
ENAMEN30001162.1GEA 0 EUR
ENAMEN30001162.1G
7-Methylisatin
7-Methylisatin
Formel: C₉H₇NO₂
Molecular Weight: 161,1574 g/mol
MDL: MFCD00022797
CAS-Nummer: 1127-59-9
EINECS: 214-431-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR