4-Amino-3-nitrobenzolsulfonamid

Lieferant: SIA ENAMINE

EN30001192.1G
ENAMEN30001192.1GEA 0 EUR
ENAMEN30001192.1G
4-Amino-3-nitrobenzolsulfonamid
4-Amino-3-nitrobenzolsulfonamid
Formel: C₆H₇N₃O₄S
Molecular Weight: 217,2000 g/mol
MDL: MFCD00116612
CAS-Nummer: 2360-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR