2,4,6-Trimethyl-N-(3-(trifluormethyl)phenyl)benzolsulfonamid

Lieferant: SIA ENAMINE

Synonyme: m-3M3FBS

EN30001199.1G
ENAMEN30001199.1GEA 0 EUR
ENAMEN30001199.1G
2,4,6-Trimethyl-N-(3-(trifluormethyl)phenyl)benzolsulfonamid
2,4,6-Trimethyl-N-(3-(trifluormethyl)phenyl)benzolsulfonamid
Formel: C₁₆H₁₆F₃NO₂S
Molecular Weight: 343,36 g/mol
MDL: MFCD00095824
CAS-Nummer: 200933-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR