2-Aminophenol-4-sulfonamid
Lieferant: SIA ENAMINE
Synonyme:
3-Amino-4-hydroxybenzenesulphonamide
EN30001211.1G
ENAMEN30001211.1GEA
0
EUR
ENAMEN30001211.1G
2-Aminophenol-4-sulfonamid
2-Aminophenol-4-sulfonamid
Formel:
C₆H₈N₂O₃S Molecular Weight: 188,2 g/mol |
MDL:
MFCD00035782 CAS-Nummer: 98-32-8 EINECS: 202-657-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...