2-Cyanphenylisocyanat

Lieferant: SIA ENAMINE

EN30001257.1G
ENAMEN30001257.1GEA 0 EUR
ENAMEN30001257.1G
2-Cyanphenylisocyanat
2-Cyanphenylisocyanat
Formel: C₈H₄N₂O
CAS-Nummer: 42066-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR