2-Cyan-4'-fluoracetanilid

Lieferant: SIA ENAMINE

EN30001284.1G
ENAMEN30001284.1GEA 0 EUR
ENAMEN30001284.1G
2-Cyan-4'-fluoracetanilid
2-Cyan-4'-fluoracetanilid
Formel: C₉H₇FN₂O
MDL: MFCD00276110
CAS-Nummer: 1735-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR