(2-Nitrophenyl)acetonitril
Lieferant: SIA ENAMINE
Synonyme:
2-Nitrobenzyl cyanide
EN30001351.1G
ENAMEN30001351.1GEA
0
EUR
ENAMEN30001351.1G
(2-Nitrophenyl)acetonitril
(2-Nitrophenyl)acetonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Siedepunkt: 304 °C (1013 hPa) Schmelzpunkt: 81…83 °C |
MDL:
MFCD00007183 CAS-Nummer: 610-66-2 EINECS: 210-231-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...