2-Methyl-4-phenylthiazol

Lieferant: SIA ENAMINE

Synonyme: 2-Methyl-4-phenylthiazole

EN30001404.1G
ENAMEN30001404.1GEA 0 EUR
ENAMEN30001404.1G
2-Methyl-4-phenylthiazol
2-Methyl-4-phenylthiazol
Formel: C₁₀H₉NS
Molecular Weight: 175,25 g/mol
MDL: MFCD00489998
CAS-Nummer: 1826-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR