2-Chlor-7-methoxy-4-methylchinolin
Lieferant: SIA ENAMINE
Synonyme:
7-Methoxy-2-chloro-4-methylquinoline
EN30001505.1G
ENAMEN30001505.1GEA
0
EUR
ENAMEN30001505.1G
2-Chlor-7-methoxy-4-methylchinolin
2-Chloro-7-methoxy-4-methylquinoline
Formel:
C₁₁H₁₀ClNO Molecular Weight: 207,66 g/mol |
MDL:
MFCD00711757 CAS-Nummer: 97892-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...